Anti-C14orf143 Mouse Monoclonal Antibody [clone: 3B3]
Lieferant: Abnova
ABNOH00090141-M06EA 394 EUR
ABNOH00090141-M06
Anti-C14orf143 Mouse Monoclonal Antibody [clone: 3B3]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant C14orf143.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: C14orf143
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen