Anti-ATG4B Rabbit Polyclonal Antibody

Lieferant: Biosensis
BSENR-157-100EA 302 EUR
BSENR-157-100
Anti-ATG4B Rabbit Polyclonal Antibody
Antikörper
FUNCTION: Cysteine protease required for autophagy, which cleaves the C-terminal part of either MAP1LC3, GABARAPL2 or GABARAP, allowing the liberation of form I. A subpopulation of form I is subsequently converted to a smaller form (form II). Form II, with a revealed C-terminal glycine, is considered to be the phosphatidylethanolamine (PE)-conjugated form, and has the capacity for the binding to autophagosomes. ENZYME REGULATION: Inhibited by N-ethylmaleimide. SUBCELLULAR LOCATION: Cytoplasm (Probable). ALTERNATIVE PRODUCTS: 5 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: Mainly expressed in the skeletal muscle, followed by brain, heart, liver and pancreas. SIMILARITY: Belongs to the peptidase C54 family.

IHC, WB and ELISA confirmed the specificity for ATG4B.
Application Information:
IHC, immunofluorescence, WB. A dilution of 1:100 to 1:2000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ATG4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminus of Human ATG4B protein
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR