Anti-Synphilin-1 Rabbit Polyclonal Antibody
Lieferant: Biosensis
BSENR-116-100EA 302 EUR
BSENR-116-100
Anti-Synphilin-1 Rabbit Polyclonal Antibody
Antikörper
Synuclein alpha interacting protein (Synphilin-1) contains several protein-protein interaction domains and interacts with alpha synuclein in neurons. Mutations of SNCAIP have been linked to Parkinson disease. The amino acid sequence of synphilin-1 shares a high level of identity with its human counterpart, particularly in regions containing ankyrin-like motifs and the coiled-coil domain. Expression pattern of synphilin-1 in tissues is similar in both mouse and human. Synphilin-1 has an important role in the formation of aggregates and cytotoxicity in Parkinson disease and also Dorfin may be involved in the pathogenic process by ubiquitylation of synphilin-1.
A high level of specificity has been shown for this antiserum by IHC on human brain.
Application Information:
IHC. A dilution of 1: 1000 to 1:2000 is recommended for this application. This antiserum will stain Lewy bodies in Dorsal raphe nucleus of Parkinson's diseased human brain. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Synphilin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human