Anti-GAPDH Rabbit Polyclonal Antibody

Lieferant: AbFrontier
ABFRLF-PA0018EA 267 EUR
ABFRLF-PA0018
Anti-GAPDH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GAPDH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR