Anti-CD46 Rabbit Polyclonal Antibody

Lieferant: Boster
BSBTPB9486EA 579 EUR
BSBTPB9486
Anti-CD46 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Membrane cofactor protein(CD46) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: CD46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR