Anti-CD55 Rabbit Polyclonal Antibody

Lieferant: Boster
BSBTPB9487EA 550 EUR
BSBTPB9487
Anti-CD55 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Complement decay-accelerating factor(CD55) detection. Tested with WB,IHC-P in Human;Mouse.

Type: Primary
Antigen: CD55
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR