Anti-TMEM173 Rabbit Polyclonal Antibody

Lieferant: Boster
BSBTPB9513EA 550 EUR
BSBTPB9513
Anti-TMEM173 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Stimulator of interferon genes protein (TMEM173) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: TMEM173
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR