Carriers for solar cells or other square devices
Lieferant: Entegris
ENTEA193-55M-0515EA 0 EUR
ENTEA193-55M-0515
Carriers for solar cells or other square devices
Wafer-Handling-Zubehör Wafer-Carrier
For handling solar cells or other square devices.
  • Pocket spacing: 4,76 mm
  • Pocket flat: 1,52 mm
  • Capacity: 25
  • Lead time: eight weeks