Anti-TIPIN Polyclonal Antibody Pair
Lieferant: Abnova
ABNOH00054962-AP21EA 835 EUR
ABNOH00054962-AP21
Anti-TIPIN Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human TIPIN.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-TIPIN (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-TIPIN (20 µg);
Type:
Antigen: TIPIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human