Anti-DNMT1 Antibody Pair

Lieferant: Abnova
ABNODP0002EA 884 EUR
ABNODP0002
Anti-DNMT1 Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the DNMT1 protein, and the other against the specific Y399 phosphorylated site of DNMT1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-DNMT1 Y399 Rabbit Polyclonal Antibody (20 µl) In PBS (0.09% (w/v) sodium azide) 2. DNMT1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: DNMT1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR