Anti-PDGFRA Antibody Pair

Lieferant: Abnova
ABNODP0010EA 884 EUR
ABNODP0010
Anti-PDGFRA Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the PDGFRA protein, and the other against the specific Y1018 phosphorylated site of PDGFRA for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-PDGFRA Y1018 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. PDGFRA Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PDGFRA
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR