Anti-FGFR1 Antibody Pair

Lieferant: Abnova
ABNODP0025EA 884 EUR
ABNODP0025
Anti-FGFR1 Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the FGFR1 protein, and the other against the specific Y463 phosphorylated site of FGFR1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-FGFR1 Y463 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. FGFR1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: FGFR1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR