Anti-GAB1 Antibody Pair

Lieferant: Abnova
ABNODP0059EA 884 EUR
ABNODP0059
Anti-GAB1 Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the GAB1 protein, and the other against the specific Y659 phosphorylated site of GAB1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-GAB1 Y659 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. GAB1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: GAB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR