Anti-RAB17 Polyclonal Antibody Pair
Lieferant: Abnova
ABNOH00064284-AP21EA 835 EUR
ABNOH00064284-AP21
Anti-RAB17 Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human RAB17.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-RAB17 (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-RAB17 (40 µl);
Type:
Antigen: RAB17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human