Anti-RAF1 Antibody Pair

Lieferant: Abnova
ABNODP0129EA 884 EUR
ABNODP0129
Anti-RAF1 Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the RAF1 protein, and the other against the specific S259 phosphorylated site of RAF1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-RAF1 S259 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. RAF1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: RAF1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR