Anti-NFKB1 Antibody Pair

Lieferant: Abnova
ABNODP0134EA 884 EUR
ABNODP0134
Anti-NFKB1 Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the NFKB1 protein, and the other against the specific S337 phosphorylated site of NFKB1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-NFKB1 S337 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. NFKB1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: NFKB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR