Anti-TYK2 Antibody Pair

Lieferant: Abnova
ABNODP0182EA 884 EUR
ABNODP0182
Anti-TYK2 Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the TYK2 protein, and the other against the specific Y1054 phosphorylated site of TYK2 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-TYK2 Y1054 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. TYK2 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: TYK2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR