Anti-PTK2B Antibody Pair

Lieferant: Abnova
ABNODP0206EA 884 EUR
ABNODP0206
Anti-PTK2B Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the PTK2B protein, and the other against the specific Y402 phosphorylated site of PTK2B for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-PTK2B Y402 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. PTK2B Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PTK2B
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR