Anti-MAPK14 Antibody Pair

Lieferant: Abnova
ABNODP0214EA 884 EUR
ABNODP0214
Anti-MAPK14 Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the MAPK14 protein, and the other against the specific T180 phosphorylated site of MAPK14 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-MAPK14 T180 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. MAPK14 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: MAPK14
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR