Anti-CREB1 Antibody Pair

Lieferant: Abnova
ABNODP0222EA 884 EUR
ABNODP0222
Anti-CREB1 Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the CREB1 protein, and the other against the specific S129 phosphorylated site of CREB1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-CREB1 S129 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. CREB1 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: CREB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR