Anti-NEIL1 Polyclonal Antibody Pair
Lieferant: Abnova
ABNOH00079661-AP21EA 835 EUR
ABNOH00079661-AP21
Anti-NEIL1 Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human NEIL1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-NEIL1 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-NEIL1 (20 µg);
Type:
Antigen: NEIL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human