Anti-F7 + F3 Antibody Pair

Lieferant: Abnova
ABNODI0178EA 794 EUR
ABNODI0178
Anti-F7 + F3 Antibody Pair
Antikörper
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the F7 protein, and the other against the F3 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. F7 Rabbit purified Polyclonal Antibody (20 µg) 2. F3 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: F7 + F3
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR