Anti-PPP2R3B Polyclonal Antibody Pair

Lieferant: Abnova
ABNOH00028227-AP22EA 835 EUR
ABNOH00028227-AP22
Anti-PPP2R3B Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human PPP2R3B.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-PPP2R3B (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-PPP2R3B (20 µg);

Type:
Antigen: PPP2R3B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR