Anti-PLCG2 + PLCG2 Polyclonal Antibody Pair

Lieferant: Abnova
ABNODP0295EA 884 EUR
ABNODP0295
Anti-PLCG2 + PLCG2 Polyclonal Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the PLCG2 protein, and the other against the specific Y1217 phosphorylated site of PLCG2 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-PLCG2 Y1217 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. PLCG2 Mouse purified Polyclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PLCG2 + PLCG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR