Anti-CDK2 + CDKN1B Antibody Pair

Lieferant: Abnova
ABNODI0272EA 794 EUR
ABNODI0272
Anti-CDK2 + CDKN1B Antibody Pair
Antikörper
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CDK2 protein, and the other against the CDKN1B protein for use in Proximity Ligation Assay

Antibody pair set content: 1. CDK2 Rabbit purified Polyclonal Antibody (20 µg) 2. CDKN1B Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: CDK2 + CDKN1B
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR