Anti-TNFRSF17 Mouse Monoclonal Antibody (Biotin)
Lieferant: Abnova
ABNOH00000608-AP42EA 884 EUR
ABNOH00000608-AP42
Anti-TNFRSF17 Mouse Monoclonal Antibody (Biotin)
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human TNFRSF17.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-TNFRSF17, IgG1 kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-TNFRSF17, IgG1 kappa (50 ?g);
Type:
Antigen: TNFRSF17
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human