Anti-CDKN1A + SKP2 Polyclonal Antibody Pair

Lieferant: Abnova
ABNODI0568EA 794 EUR
ABNODI0568
Anti-CDKN1A + SKP2 Polyclonal Antibody Pair
Antikörper
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CDKN1A protein, and the other against the SKP2 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. CDKN1A Rabbit purified Polyclonal Antibody (20 µg) 2. SKP2 Mouse purified Polyclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: CDKN1A + SKP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR