Anti-PNMA2 Mouse Monoclonal Antibody (Biotin)
Lieferant: Abnova
ABNOH00010687-AP43EA 835 EUR
ABNOH00010687-AP43
Anti-PNMA2 Mouse Monoclonal Antibody (Biotin)
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human PNMA2.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-PNMA2 (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-PNMA2, IgG2b kappa (50 ?g);
Type:
Antigen: PNMA2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human