Anti-RALB Antibody Pair

Lieferant: Abnova
ABNOH00005899-AP61EA 835 EUR
ABNOH00005899-AP61
Anti-RALB Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human RALB.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-RALB (100 µg); 2. Detection Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-RALB (50 µg);

Type:
Antigen: RALB
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR