Anti-APBB1 Polyclonal Antibody Pair

Lieferant: Abnova
ABNODP0056EA 884 EUR
ABNODP0056
Anti-APBB1 Polyclonal Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the APBB1 protein, and the other against the specific S175 phosphorylated site of APBB1 for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-APBB1 S175 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. APBB1 Mouse purified Polyclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: APBB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR