Anti-S100 A8 Antibody Pair

Lieferant: Abnova
ABNOH00006279-AP52EA 835 EUR
ABNOH00006279-AP52
Anti-S100 A8 Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human S100A8.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-S100A8 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-S100A8 (50 µg);

Type:
Antigen: S100 A8
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR