Anti-TCF7 Antibody Pair

Lieferant: Abnova
ABNOH00006932-AP11EA 3140 EUR
ABNOH00006932-AP11
Anti-TCF7 Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human TCF7.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-TCF7 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-TCF7, IgG2a kappa (20 µg);

Type:
Antigen: TCF7
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR