Anti-ITIH1 Polyclonal Antibody Pair
Lieferant: Abnova
ABNOH00003697-PW1EA 835 EUR
ABNOH00003697-PW1
Anti-ITIH1 Polyclonal Antibody Pair
Antikörper
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-ITIH1 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-ITIH1 (50 µg)
Type:
Antigen: ITIH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human