Anti-GLRX3 Polyclonal Antibody Pair

Lieferant: Abnova
ABNOH00010539-AP22EA 835 EUR
ABNOH00010539-AP22
Anti-GLRX3 Polyclonal Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human GLRX3.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-GLRX3 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-GLRX3 (20 µg);

Type:
Antigen: GLRX3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR