Anti-CCL15 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00006359-PW1EA 835 EUR
ABNOH00006359-PW1
Anti-CCL15 Rabbit Polyclonal Antibody
Antikörper
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

  • Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-CCL15 (300 μl); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-CCL15 (50 μl)

Type:
Antigen: CCL15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR