Anti-CCL2 Mouse Monoclonal Antibody (Biotin)

Lieferant: Abnova
ABNOH00006347-AP43EA 835 EUR
ABNOH00006347-AP43
Anti-CCL2 Mouse Monoclonal Antibody (Biotin)
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human CCL2.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CCL2 (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-CCL2, IgG2a kappa (50 ?g);

Type:
Antigen: CCL2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR