Anti-CCL2 Mouse Monoclonal Antibody (Biotin)
Lieferant: Abnova
ABNOH00006347-AP44EA 835 EUR
ABNOH00006347-AP44
Anti-CCL2 Mouse Monoclonal Antibody (Biotin)
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human CCL2.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-CCL2 (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-CCL2, IgG2a kappa (50 ?g);
Type:
Antigen: CCL2
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human