Anti-Dectin-1 Rabbit Polyclonal Antibody
Lieferant: BioVision
BIOV3891-100EA 288 EUR
BIOV3891-100
Anti-Dectin-1 Rabbit Polyclonal Antibody
Dectin-1also known as beta-glucan receptor is a small type II transmembrane receptor containing lectin-like carbohydrate recognition domain. Dectin-1 plays a role in beta-glucan dependant, nonopsonic recognition of zymosan by primary macropahge. Dectin-1 is expressed on macrophages and neutrophils and on some dendritic cells subpopulation.
Recommended Dilutions: Western Blot: 0.5-4 µg/ml
Type: Primary
Antigen: Dectin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat