Meine Sitzung geöffnet lassen?
Endet in 
Ihre Sitzung ist abgelaufen
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Anti-PCAF Rabbit Polyclonal Antibody

Lieferant: BioVision
The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. P300/CBP-associated factor (PCAF) is a transcriptional coactivator that works both as a histone lysine acetyltransferase, through its HAT domain, and as an acetyl-lysine reader through its conserved bromodomain located directly C-terminal to the HAT domain. The PCAF bromodomain binds acetylated histone H3 and H4 as well as non-histone targets. Bromodomain binding is dictated by the position of the acetylated lysine as well as interactions with specific residues flanking the acetyl-lysine. PCAF also specifically binds the HIV viral protein Tat on acetylated K50 to regulate its transactivating activity and help induce chromatin remodeling of proviral genes, thereby promoting transcription of viral proteins.

Recommended Dilutions: Western Blot: ~1:1000.

Type: Primary
Antigen: PCAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human. Predicted Cross Reactivity With Mouse Samples.



 

BESTELLEN

Erweitert / Kompakt
Größe VE Packung Katalognummer Menge
100 µl 100 µl Vial BIOV6763-100
Generate Barcodes Barcode Label Format: 
PDF Katalogseiten erstellen

TECHNISCHE DATEN

Antigen symbol PCAF
Reaktivität Human, Mouse
Konjugation Unconjugated
Klonalität Polyclonal
Antikörper Typ Primary
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms KAT2B
Epitope C-terminal
Cross adsorption No
Immunogen This PCAF antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 802-832 amino acids from the C-terminal region of human PCAF.
Purification This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.
Form Liquid
Storage buffer PBS with 0,09% (W/V) sodium azide
Lagertemperatur – 20 °C
Transporttemperatur 4 °C
Enter Details

Wählen Sie den Artikel dessen Zertifikat Sie gesucht haben

Chargennummer ist ein Pflichtfeld. Falls Sie diese nicht zur Verfügung haben, tragen Sie "x" für VWR Chemikalien ein.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge