Anti-FGF-18 Rabbit Polyclonal Antibody
Lieferant: BioVision
BIOV5082-100EA 276 EUR
BIOV5082-100
Anti-FGF-18 Rabbit Polyclonal Antibody
The FGFs plays an important role in regulating tissue growth, morphogenesis and tissue repair. FGF-18 is a newly discovered heparin binding growth factor that stimulates the proliferation and activation of cells that express FGF receptors.
Recommended Dilutions: Western Blot: 0.5-4 µg/ml
Type: Primary
Antigen: FGF-18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human