Anti-GDNF Rabbit Polyclonal Antibody
Lieferant: BioVision
BIOV5098-100EA 384 EUR
BIOV5098-100
Anti-GDNF Rabbit Polyclonal Antibody
Glial cell line-derived neurotrophic factor (GDNF) has been identified as a potent neurotrophic factor that enhances survival of midbrain dopaminergic neurons. GDNF is a glycosylated, disulfide-bonded homodimer. It promotes the survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake. On the basis of these findings, it has been sμggested that GDNF may play a role in the treatment of Parkinson’s disease, which is marked by progressive degeneration of midbrain dopaminergic neurons.
Recommended Dilutions: Western Blot: 0.5-4 µg/ml
Type: Primary
Antigen: GDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat