Human Recombinant sIL-2Ralpha
Lieferant: Tonbo Biosciences
21-8702-U005EA 70.4 EUR
TNBO21-8702-U005 TNBO21-8702-U500 TNBO21-8702-U100 TNBO21-8702-U025
Human Recombinant sIL-2Ralpha
The high-affinity IL-2 receptor is a trimeric complex made up of IL-2R alpha (CD25), IL-2R beta (CD122) and the common gamma chain (IL-2R gamma, CD132). The alpha chain is cleaved from the cell membrane through proteolytic processing to release soluble IL-2R alpha. IL-2R alpha binds IL-2 with low affinity and is not essential for IL-2 signaling, but when combined with the beta and gamma chains it greatly increases affinity for IL-2. IL-2R alpha is expressed during early T and B lymphocyte development and is also induced during mature T and B cell activation. It has been reported to enhance Th17 responses in vitro.