Media bottles with cap, round
Lieferant: Hach
HACH2495012EA 0 EUR
HACH2495012
Media bottles with cap, round
Flaschen Zellkulturflaschen
PS, transparent
  • Non-fluorescent
  • Break-resistant
  • Presterilised
  • Have 100 ml fill-to line
These disposable bottles are used with medium containing MUG or other fluorescing reagent. Works well with P-A Broth ampules and PathoScreen™ medium.