Anti-BLVRA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP20430CEA 348 EUR
ABGEAP20430C
Anti-BLVRA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BLVRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR