N-Boc-2-pyrroleboronic acid 96%
|
Achtung
|
H53078.03EA
137
EUR
H53078.03
H53078.06
N-Boc-2-pyrroleboronic acid 96%
N-Boc-2-pyrroleboronic acid
Formel:
C₉H₁₄BNO₄ Molekulargewicht: 211,02 g/mol Schmelzpunkt: 105…108 °C |
MDL Number:
MFCD01318939 CAS-Nummer: 135884-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...