1-tert-Butyl-3-ethylpyrrolidine-1,3-dicarboxylate 97%
|
Achtung
|
H57430.03EA
83.6
EUR
H57430.03
1-tert-Butyl-3-ethylpyrrolidine-1,3-dicarboxylate 97%
1-tert-Butyl-3-ethylpyrrolidine-1,3-dicarboxylate
Formel:
C₁₂H₂₁NO₄ Molekulargewicht: 243,30 g/mol |
MDL Number:
MFCD03791256 CAS-Nummer: 170844-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...