4-Hexylphenylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
H64555.03EA
207
EUR
H64555.03
4-Hexylphenylboronsäure 97%
4-Hexylphenylboronsäure
Formel:
C₁₂H₁₉BO₂ Molekulargewicht: 206,09 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04039029 CAS-Nummer: 105365-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...