Dihydrofluorescein diacetate 97%

Lieferant: Thermo Scientific

J65759.03EA 147 EUR
J65759.03
Dihydrofluorescein diacetate 97%
Dihydrofluorescein diacetate

Applications: Fluorescent dye for detecting intracellular oxidants

Formel: C₂₄H₁₈O₇
Schmelzpunkt: 213…215 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00005056
CAS-Nummer: 35340-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR