Pentadecan-15-olid

Synonyme: Oxa-2-cyclohexadecanonω-Pentadecalacton15-HydroxypentadecansäurelactonOxacyclohexadecan-2-on

SPCMC2800-25KGBLEA 0 EUR
SPCMC2800-25KGBL
Pentadecan-15-olid
Pentadecan-15-olid
Formel: C₁₅H₂₈O₂
Molekulargewicht: 240,39 g/mol
MDL Number: MFCD00039667
CAS-Nummer: 106-02-5
EINECS: 203-354-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR