Process supplement, HyClone Cell Boost™ 4
Lieferant: HyClone products (Cytiva)
HYCLSH30857.01EA 248 EUR
HYCLSH30857.01
Process supplement, HyClone Cell Boost™ 4
Zellkultur-Ergänzungen und -Zusätze
HyClone™ Cell Boost™ 4 Supplement is designed to provide nutrients such as amino acids, vitamins, glucose, trace elements, and growth factors as part of fed-batch cell culture strategies.
  • Chemically defined, protein-free and free of animal-derived components (ADCF)
  • Tested on a variety of cell lines including CHO
  • Ideal for supplementing nutrients for bioprocessing and media customisation applications
  • Manufactured according to cGMP guidelines